Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Synthetic LKBtide | Avantor

Synthetic LKBtide

1 MG
Catalog # 77559-280
Supplier:   Sino Biological
Fournisseur #:  L15-58
Synthetic LKBtide
Catalog # 77559-280
Supplier:  Sino Biological
CAS Number:  
Synthetic LKBtide

1 MG

CAS Number:  
Price
$136.11
Log in pour la disponibilité
Each(1/Itm/Bx)
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

The synthetic peptide LKBtide (LSNLYHQGKFLQTFCGSPLY to RRRC) is derived from Human NUAK2 (amino acid 196 to 215) and can be phosphorylated by protein Serine/Threonine kinase 11 (STK11), also known as LKB1.

Spécifications

Spécifications

  • Taille:
    1 MG
  • Cat. No.:
    77559-280
  • Protein/peptide type:
    Synthetic
  • Protein/peptide name:
    LKBtide
  • Storage conditions:
    Store product at –20 °C. For optimal storage, aliquot diluted product into smaller quantities and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
  • Formulation:
    1 mg of peptide supplied as a lyophilized powder.
  • Conjugation:
    Unconjugated
  • Tested applications:
    Kinase Assay
  • Molecular weight:
    2785.24
  • Shipping temperature:
    Substrates are provided as liquid or lyophilized powder. They are recommended to be shipped out with blue ice.

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.