Complete Tissue Embedding Center
Catalogue # 77435-284
Fournisseur: PRECISIONARY INSTRUMENTS LLC
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Description:Complete tissue embedding center
- Cat. no.:77435-284
Spécifications
À propos de cet article
Paraffin wax embedding stations for tissue to section on the rotary microtome.
- To embed samples in paraffin wax
Embedding stations – also called tissue processors, tissue stations, embedding machines and tissue embedding centers (TECs) – embed paraffin into tissue specimens. HOT and COLD plates included.