ContactUsLinkComponent
Tissue Floating Bath
  77435-368
undefined
Tissue Floating Bath
  77435-368
 :  TF1000
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Tissue floating bath
  • Cat. no.:
    77435-368
The precisionary instruments tissue floating bath is a reliable and efficient solution for softening embedded tissue specimens without causing wrinkles or distortions. With a teflon-coated chamber for easy cleaning, a low-profile interior chamber to reduce glare.
  • The tissue floating bath is used after sectioning with a rotary microtome
For use after sectioning paraffin wax.