QuEChERS Dispersive Solid Phase Extraction (SPE) Kits
QuEChERS Dispersive Solid Phase Extraction (SPE) Kits
QuEChERS Dispersive Solid Phase Extraction (SPE) Kits
Certificates
QuEChERS Dispersive Solid Phase Extraction (SPE) Kits
4 Options Available Below
Product Details & Documents
QuEChERS dispersive solid phase extraction (SPE) kits supplied in 15 ml centrifuge tubes accommodate the aliquot volumes specified by current AOAC and EN methodologies.
  • Non hazardous
The sorbent extracts the sample matrix, while MgSO₄ helps remove excess water and improve analyte partitioning.
Product Family Options
Informations sur le produit
  • Désignation
    Capacité
    Availability
    Price
  • QuEChERS dSPE kit, TFDA method, includes PSA, MgSO₄
    15 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $362.00
    $362.00
    Each(1/Itm/Bx)
       $362.00
    Supplier Number: 5982-6662
  • QuEChERS dSPE kit, TFDA method, includes PSA, C18, MgSO₄
    15 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $380.00
    $380.00
    Each(1/Itm/Bx)
       $380.00
    Supplier Number: 5982-6663
  • QuEChERS dSPE kit, TFDA method, includes PSA, C18, GCB, MgSO₄
    15 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $479.00
    $479.00
    Each(1/Itm/Bx)
       $479.00
    Supplier Number: 5982-6664
  • QuEChERS dSPE kit, TFDA method, includes animal origin
    15 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $467.00
    $467.00
    Each(1/Itm/Bx)
       $467.00
    Supplier Number: 5982-6665