Ward's® Aluminum Weighing Boats
New Product
Boats for general-purpose weighing, dispensing, storage, food processing, evaporating, or drying.
- Sturdy yet lightweight boats feature a smooth, flat bottom for sample handling and a wall for rigidity
- Finger tab handle can be etched for identification prior to weighing
- Disposable
- Available in a wide range of sizes
- All sizes come in packs of 100
Versatile weighing boats catering to a broad spectrum of tasks.
Frequently Bought Together