ASI™ Supreme Plain Surface Non-Charged Microscope Slides, Frosted White Writing Area
Catalogue # 104028-348
Fournisseur: Alkali Scientific
Spécifications
- Description:ASI™ Supreme plain surface non-charged microscope slides, frosted white writing area
- Cat. no.:104028-348
Spécifications
À propos de cet article
Plain surface non-charged microscope slides are 75×25 mm and 1.0 mm thick. Their key feature is the frosted end on one side, which makes it easier to label them with pencils or pens than on other microscope glasses, as it is sand-blasted glass and not etched.
- Frosted white writing area
- Non-charged
- Glass
Microscope slides are suitable for most type of laboratory research and allow all type of mounting methods, from dry mounting to wet or temporary mounting, prepared mounting and strew mounting. Typically a sample is mounted or placed on the slide and then both are put under the microscope for viewing.
Packaging: 20 inner packs of 72, 1440/case.