ContactUsLinkComponent
Radiant™ DNA Extract and Amplify Tissue PCR Kit
Radiant™ DNA Extract and Amplify Tissue PCR Kit
Catalog # 104026-574
Supplier:  Alkali Scientific
CAS Number:  
undefined
Radiant™ DNA Extract and Amplify Tissue PCR Kit
Catalog # 104026-574
Supplier:  Alkali Scientific
Supplier Number:  C462
CAS Number:  
Specifications
  • Description:
    Radiant™ Extract and amplify tissue PCR kit
  • Size:
    80 Reactions, (2×1 ml 2X HS Taq Red, 1×1.6 ml 5X extract buffer A, 1×0.08 ml 10X Extract Buffer B)
  • Cat. no.:
    104026-574
Specifications
About this item
Effortlessly extract and amplify DNA from tissue samples with the Radiant™ DNA extract and amplify tissue PCR kit.
  • 15 minute extraction process
  • No overnight lysing/extracting
  • PCR with supplied mix
  • Red loading buffer included in PCR mix
Radiant™ Extract and amplify tissue PCR kit contains all the reagents needed to rapidly extract and amplify genomic DNA from mouse tail, ear punch and other mammalian tissues, buccal swabs, hair shafts and saliva. Speed up your Genotyping, no overnight lysing Protocol: extraction solutions, included: 15 minutes then PCR as directed with supplied Radiant™ Red 2X mix. During the quick 5 minute extraction process, DNA is released from the starting material by mixing the sample with ASI's extraction solution. This is followed by a 10 minute incubation at 95° to help inactivate proteins and genomic DNA. No overnight extraction. There is no need for mechanical disruption, organic extraction, column purification or precipitation of the DNA.
Caution: Limitations of use: This product is intended for research purposes only and is not intended for any animal or human therapeutic use.