ContactUsLinkComponent
CellPro™ Flat Bottom, Non-Treated Cell Culture Plates with Lids
Catalog # 104027-190
Supplier:  Alkali Scientific
CellPro™ Flat Bottom, Non-Treated Cell Culture Plates with Lids
Catalog # 104027-190
Supplier:  Alkali Scientific
Supplier Number:  TL5384
Specifications
  • No. of wells:
    384
  • Description:
    Cell culture plates with lids, non-treated
  • Cat. no.:
    104027-190
  • Packaging:
    50/case
  • Sterile:
    Sterile
Specifications
About this item
Cultivate cells with ease using CellPro™ flat bottom, non-treated cell culture plates, equipped with lids for secure containment of samples.
  • High clarity, 100% virgin polystyrene
  • Vacuum gas plasma tissue culture treatment
  • Clear lot number for batch tractability
  • Peel-to-open, individually wrapped and sterilized by gamma irradiation
  • Markings of well coordinates available for 12, 24, 48 and 96 well plates
  • Non-pyrogenic, DNase/RNase free
  • Gamma sterilized
CellPro™ flat bottom cell cultureplates, designed for a wide range of cell culture applications. These plates feature a flat bottom surface, providing optimal stability for cell growth and attachment. Each plate comes with a matching lid to prevent contamination and evaporation, ensuring the integrity of your cell cultures. Manufactured from high-quality materials, these non-treated plates offer versatility for use with various cell lines and culture media.
CellPro™ flat bottom cell culture plates are used for performing routine cell culture experiments or specialized assays with consistent and reproducible results.