Specifications
- Color:Blue
- Description:Replacement caps, standard, 10/pack
- Cat. no.:104026-954
- Thread Size:GL 32
Specifications
About this item
Keep your Hybex™ media storage bottles sealed with replacement caps for Hybexª media storage bottle standard blue (GL 32), available in a pack of 10.
- Replacement cap for Hybexª media storage bottles
- Standard blue (GL 32)
Ensure the integrity of your Hybex™ media storage bottles with the replacement caps, designed specifically for the standard blue GL 32 bottle. This pack of ten replacement caps provides a reliable solution for maintaining a secure seal, preserving the contents of your media storage bottles. Crafted with precision and durability in mind, these caps offer a snug fit to prevent leaks and contamination, safeguarding valuable samples. With easy installation and robust construction, the replacement caps for Hybex™ media storage bottles offer peace of mind and confidence in your laboratory operations, ensuring optimal storage conditions for media and reagents.