Radiant™ Probe 1-Step Lo-Roxx qPCR Virus Kit
Catalogue # 104026-642
Fournisseur: Alkali Scientific
Numéro CAS:
Spécifications
- Description:Radiant™ probe 1-step Lo-ROX qPCR virus kit, 4X
- Cat. no.:104026-642
Spécifications
À propos de cet article
Detect viral targets efficiently with Radiant™ Probe 1-Step Lo-Roxx qPCR Virus kit, designed for probe-based quantitative PCR.
- Recommended kit for SARS-CoV-2 diagnostics
- Validated for qualitative detection of SARS-CoV-2 nucleic acid
- Concentrated 4X mix format, ideal for high-throughput, highly multiplexed assays
- Ultra-sensitive detection of RNA viruses
- Earlier detection of a wide range of template amounts
- Includes UltraScript RTase for thermostable reverse transcription up to 55 °C
- Advanced RNase inhibitor
- Antibody-mediated hot start technology
- Compatible with all real-time PCR platforms - standard and fast cycling conditions
cDNA Synthesis and qPCR in a single tube. Highly concentrated 4X mix allows increased sample and viral input. This optimizes sample sensitivity with small reaction volumes. Engineered for use with a wide range of probe technologies including TaqMan®, Scorpions® and molecular beacon probes.
Mix includes the thermostable Radiant™ Reverse transcriptase which is blended with an advanced RNase inhibitor to prevent degradation of RNA by contaminating RNase.