ContactUsLinkComponent
Radiant™ cDNA Synthesis Kit, 1-Step
  104027-918
 :  
undefined
Radiant™ cDNA Synthesis Kit, 1-Step
  104027-918
 :  QC1125
 :  
  • Description:
    Radiant™ cDNA synthesis kit, 1-Step, 25 reactions, (1×0.1 ml 5X cDNA mix, 1×0.025 ml 20X RTase)
  • Cat. no.:
    104027-918
Conduct efficient cDNA synthesis with Radiant™ cDNA synthesis kit, offering a convenient 1-step process.
  • Unbiased representation of 5’ and 3’ mRNA transcript ends
  • Sensitive detection of low copy number transcripts
  • High cDNA yields from as little as 4pg total RNA
  • Simple 2 tube system
  • Supplied with a proprietary ratio of anchored oligo dT and random hexamers
The Radiant™ cDNA Synthesis kit uses the latest developments in reverse transcriptase technology and buffer chemistry to enhance cDNA synthesis speed and yield with accurate transcript representation. The reverse transcriptase, buffer system and combination of random hexamers with anchored oligo(dT) allow for unbiased, efficient, sensitive cDNA synthesis.
High quality cDNA synthesis for downstream qPCR analysis is essential for successful expression studies. Many factors affect cDNA synthesis including the reverse transcriptase, buffer systems, enhancers and priming strategy. The Radiant™ cDNA synthesis mix removes the need for user optimization of these critical factors.
High quality cDNA synthesis for downstream qPCR analysis is essential for successful expression studies. Many factors affect cDNA synthesis including the reverse transcriptase, buffer systems, enhancers and priming strategy. The Radiant™ cDNA synthesis mix removes the need for user optimization of these critical factors.
The modified MmlV reverse transcriptase (RTase) is both thermo stable and extremely active. The enzyme is blended with RNase inhibitor preventing degradation of RNA by contaminating RNase.
The RTase is not inhibited by ribosomal and transfer RNAs, total RNA is an ideal substrate. The 5X cDNA synthesis mix can be used with up to 4.0 µg total RNA. The relative concentrations of random hexamers and anchored oligo (dT) have been optimized for the generation of cDNA for use in real-time PCR experiments.