Order Entry
Canada
ContactUsLinkComponent
ProCycle™ 384 Well ABI®-Type PCR Plates, Clear, Full Skirt Edge, A24 notch
ProCycle™ 384 Well ABI®-Type PCR Plates, Clear, Full Skirt Edge, A24 notch
Catalog # 104026-758
Supplier:  Alkali Scientific
undefined
ProCycle™ 384 Well ABI®-Type PCR Plates, Clear, Full Skirt Edge, A24 notch
Catalog # 104026-758
Supplier:  Alkali Scientific
Supplier Number:  PC384

Specifications

  • No. of wells:
    384
  • Description:
    PCR plates, full skirt edge, A24notch, clear, 10/pack
  • Cat. no.:
    104026-758

Specifications

About this item

Perform high-throughput PCR assays with ProCycle™ 384 Well ABI®-type PCR plates, featuring a clear design and full skirt edge, available in packs of 10.

  • Universal compatibility: PCR plate is compatible with a broad range of thermal cyclers, ensuring versatility and adaptability in your laboratory
  • Easy well identification: High-contrast, black alphanumeric indexing facilitates quick and accurate well identification, streamlining workflow
  • Optimal heat transfer: Ultra-thin, uniform wells ensure optimal heat transfer, promoting high reaction efficiency and reproducibility
  • Sample protection: Well ridges help reduce sample evaporation when sealing with film, foil or strip caps, preserving sample integrity
  • High clarity, ultra-pure material: PCR plate is manufactured from high-clarity, ultra-pure polypropylene, meeting FDA 21 CFR 177.1520 and USP Class VI standards
  • Purity assurance: The plate is certified free of detectable DNase, RNase, DNA, PCR inhibitors and has been tested to be pyrogen-free

ProCycle™ 384 Well ABI®-Type PCR plates are meticulously engineered for high-throughput PCR applications. These clear plates feature a full skirt edge design with an A24 notch, ensuring compatibility with leading thermal cyclers. With a well-optimized design, these plates provide maximum thermal conductivity for efficient PCR amplification, delivering reliable and reproducible results. Each pack contains 10 plates, making them an ideal choice for laboratories conducting large-scale PCR experiments. ProCycle™ PCR plates for exceptional performance and consistency in your molecular biology workflows.

PC384 Compatible with the following machines (384 well head/block upgrades):
• ABI 7700
• Life Technologies/ThermoFisher Quantstudio 6 Pro and 7 and Quantstudio 12K FLEX