Immunoplates, 96 well, ELISA Maxibinding, Polystrene, Flat Bottom with Lid
Catalogue # 104028-404
Fournisseur: Alkali Scientific
Spécifications
- Color:Clear
- Description:96 well microplate, maxibinding, F-type
- Material:PS
- Size:85.4×127.6 mm
- Cat. no.:104028-404
- Packaging:10/sleeve, 100/case
Spécifications
À propos de cet article
Perform ELISA assays efficiently with Immunoplates, featuring a clear PS construction, 96 wells, flat bottom with lid, available in cases of 100.
- Clear polystyrene
- Surface chemistry for optimal binding is hydrophilic and ideal for protein research and antibody sandwich assays
- Alphanumerically labeled rows and columns
Maximize the efficiency of ELISA assays with the Immunoplates. Constructed from clear polystyrene, these 96-well plates feature a flat bottom design and come with lids for convenient sample protection. The ELISA MAXIBINDING surface ensures optimal antibody binding and assay sensitivity. Each case contains 100 plates, providing a dependable solution for high-throughput ELISA applications. Trust in the superior quality and performance of these immunoplates to streamline your laboratory workflows and achieve accurate results.