Round Bottom Flasks
Supplier: Heidolph NA, LLC
Jump to:
À propos de cet article
These round bottom flasks are made up of glass, for use with StarFish™ multi-experiment heating and stirring work station.
16 Options Available Below
Product Details & Documents
These round bottom flasks are made up of glass, for use with StarFish™ multi-experiment heating and stirring work station.
Product Family Options
Informations sur le produit
- CapacitéST Joint No.Bottom StyleAvailabilityPrice
- 5 ml14/23RoundDiscontinued-Supplier Number: 015890700Spécifications:
More
Specifications
Cat. No.CA11007-820 - Supplier Number: 015890700CASpécifications:
More
Specifications
Cat. No.CA11009-134 - 5 ml10/19RoundDiscontinued-Supplier Number: 015890710Spécifications:
More
Specifications
Cat. No.CA11007-822 - 10 ml14/23RoundDiscontinued-Supplier Number: 015890680Spécifications:
More
Specifications
Cat. No.CA11007-816 - Supplier Number: 015890680CASpécifications:
More
Specifications
Cat. No.CA11009-130 - 10 ml10/19RoundDiscontinued-Supplier Number: 015890690Spécifications:
More
Specifications
Cat. No.CA11007-818 - Supplier Number: 015890690CASpécifications:
More
Specifications
Cat. No.CA11009-132 - 25 ml24/29RoundDiscontinued-Supplier Number: 015890660Spécifications:
More
Specifications
Cat. No.CA11007-812 - Supplier Number: 015890660CASpécifications:
More
Specifications
Cat. No.CA11009-126 - 25 ml19/26RoundDiscontinued-Supplier Number: 015890670Spécifications:
More
Specifications
Cat. No.CA11007-814 - Supplier Number: 015890670CASpécifications:
More
Specifications
Cat. No.CA11009-128 - 25 ml14/23RoundDiscontinued-Supplier Number: 015890750Spécifications:
More
Specifications
Cat. No.CA11007-830 - Supplier Number: 015890750CASpécifications:
More
Specifications
Cat. No.CA11009-144 - 50 ml24/29RoundDiscontinued-Supplier Number: 015890640Spécifications:
More
Specifications
Cat. No.CA11007-808 - Supplier Number: 015890640CASpécifications:
More
Specifications
Cat. No.CA11009-122 - 50 ml19/26RoundDiscontinued-Supplier Number: 015890650Spécifications:
More
Specifications
Cat. No.CA11007-810 - Supplier Number: 015890650CASpécifications:
More
Specifications
Cat. No.CA11009-124 - 50 ml14/23RoundDiscontinued-Supplier Number: 015890740Spécifications:
More
Specifications
Cat. No.CA11007-828 - Supplier Number: 015890740CASpécifications:
More
Specifications
Cat. No.CA11009-142 - 100 ml24/29RoundDiscontinued-Supplier Number: 015890620Spécifications:
More
Specifications
Cat. No.CA11007-804 - Supplier Number: 015890620CASpécifications:
More
Specifications
Cat. No.CA11009-118 - 100 ml19/26RoundDiscontinued-Supplier Number: 015890630Spécifications:
More
Specifications
Cat. No.CA11007-806 - Supplier Number: 015890630CASpécifications:
More
Specifications
Cat. No.CA11009-120 - 100 ml24/29RoundDiscontinued-Supplier Number: 015890730Spécifications:
More
Specifications
Cat. No.CA11007-826 - Supplier Number: 015890730CASpécifications:
More
Specifications
Cat. No.CA11009-140 - 150 ml24/29RoundDiscontinued-Supplier Number: 015890600Spécifications:
More
Specifications
Cat. No.CA11007-800 - 150 ml19/26RoundDiscontinued-Supplier Number: 015890610Spécifications:
More
Specifications
Cat. No.CA11007-802 - Supplier Number: 015890610CASpécifications:
More
Specifications
Cat. No.CA11009-116 - 250 ml24/29RoundDiscontinued-Supplier Number: 015890580Spécifications:
More
Specifications
Cat. No.CA11007-796 - Supplier Number: 015890580CASpécifications:
More
Specifications
Cat. No.CA11009-110 - 250 ml19/26RoundDiscontinued-Supplier Number: 015890590Spécifications:
More
Specifications
Cat. No.CA47800-120 - Supplier Number: 015890590CASpécifications:
More
Specifications
Cat. No.CA11009-112 - 250 ml24/29RoundDiscontinued-Supplier Number: 015890720Spécifications:
More
Specifications
Cat. No.CA11007-824 - Supplier Number: 015890720CASpécifications:
More
Specifications
Cat. No.CA11009-138