Organize and protect microscope slides with the Microscope slide box, providing secure storage for samples.
- Numbered index sheet in lid for quick identification
- Foam liner
- Stackable
- ABS construction is durable and impact-resistant
- Hinged lid
- Unique sliding compression latch
The Microscope slide box provides a secure and organized solution for storing and transporting delicate microscope slides in laboratory settings. Designed with precision and durability in mind, this box offers protection for valuable slides, preventing damage or breakage during handling and storage. With its compact and lightweight construction, it is easy to carry and conveniently fits into standard laboratory cabinets or drawers. The box is specifically designed to acco mmodate standard microscope slides, ensuring a snug fit and minimizing movement to maintain slide integrity. Whether conducting routine microscopy examinations or storing slides for long-term archival purposes, the Microscope slide box offers a reliable and efficient storage solution, helping to maintain the quality and integrity of samples.