To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Size:19 L (5 gal.)
- Cat. no.:CA89089-206
- Packaging:Plastic pail
- Supplier No.:1L1205
Specifications
About this item
LabKlenz™ 120 Detergent is a phosphate-free, liquid, alkaline detergent for use in automated laboratory glassware washing applications. It is compatible with a variety of substrates including stainless steel, mild steel, and copper. It is extremely free rinsing to meet critical laboratory requirements.
- Efficacy - Effective removal of laboratory residues in both soft and hard water
- Flexibility - Mild alkalinity for better materials compatibility
- Cost savings - Reduces utility consumption and cleaning time due with low foaming, free-rinsing formulation
- Environmentally friendly - phosphate-free formulation with biodegradable surfactants
- Reliability - Manufactured in a cGMP compliant, FDA registered and ISO 9001 certified facility
LabKlenz™ 120 Detergent is specially formulated to meet the unique cleaning and materials compatibility requirements found in laboratory glassware washing applications.