CIP Neutralizer® Alkaline Based Neutralizers
Catalog # 77531-110
Supplier: Steris
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Size:1136 L (300 gal.)
- Cat. no.:77531-110
- Packaging:Plastic tote
Specifications
About this item
CIP Neutralizer® Alkaline neutralizer is a liquid blend of sodium hydroxide and potassium hydroxide suitable for a variety of applications where raising the pH or increasing the alkalinity of a solution is required.
- Productivity - Raises the pH of solution utilizing a blend of potassium and sodium hydroxide
- Environmentally friendly - Formulated with low mercury content in order to meet environmental requirements
- Reliability - Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility
CIP Neutralizer® Alkaline based neutralizer is specially formulated for neutralization of acidic solutions.