Order Entry
Canada
ContactUsLinkComponent
Septihol® Ethyl WFI se Alcohol Solution
Catalogue # 77531-088
Fournisseur:  Steris
New Product
Septihol® Ethyl WFI se Alcohol Solution
Catalogue # 77531-088
Fournisseur:  Steris
Supplier Number:  625608WR
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Spécifications

  • Size:
    4×1 gal.
  • Cat. no.:
    77531-088
  • Packaging:
    Plastic bottle

Spécifications

À propos de cet article

Disinfectant for microbial control.

  • Efficacy - Biocidal and fungicidal EPA claims
  • Productivity - Ready to use alcohol eliminates in-house production validation and documentation concerns
  • Rinsing - Evaporates quickly with no residue
  • Reliability - Each lot is tested to ensure low endotoxins

Septihol® Ethyl WFI se Alcohol Solution provides microbial control and removes residue from hard surfaces. It can be used straight out of the container and be applied poured or sprayed. Since it contains no stabilizers or denaturants, it evaporates quickly and completely.

Septihol® Ethyl WFI se is a non sterile mixture of ethanol, isopropanol, methanol and United States Pharmacopeia (USP) Water For Injection (WFI) designed to remove organic and inorganic residues in aseptic processing areas. Its EPA biocidal and fungicidal claims are proven in presence of 5% organic soil on hard, nonporous surfaces. The formula does not contain stabilizers or denaturants, so it evaporates quickly and completely.

CAS: 64-17-5; 67-63-0; 67-56-1
UN 1170
Hazard class: 3
Packing group: II

Caution: Danger, GHS02; GHS07; GHS08.