ProKlenz® RESTORE High Performance Acid Based Cleaners
Supplier: Steris
Certificates
About this item
ProKlenz® RESTORE Cleaner has an acidic pH for the removal of type I rouge and all-purpose cleaning.
- Standardization - Globally available product enables harmonized cleaning protocols across sites
- Sustainability - Free rinsing formula minimizes utility consumption and detergent residue
- Effectiveness - An acidic pH removes type I rouge while a low foaming system supports use in high-impingement systems
- Efficiency - A low use-concentration supports cost-effective and optimized cleaning
- Quality - Manufactured within an FDA registered plant, ISO 9001:2015 and ISO 13485:2016 compliant, to meet the highest standard in manufacturing and quality assurance
ProKlenz® RESTORE High performance acid based cleaner is a globally available, low foaming acid based product for type I rouge removal and all-purpose cleaning. The global acceptability of ProKlenz® RESTORE accommodates the pharmaceutical, biotechnology, cosmetic, and nutraceutical industries by removing difficult process residues.
Frequently Bought Together