Automatic Liquid Sampler Kit
Basic automatic liquid sampler supply kit
AG07673-60840
: 07673-60840
Basic supplies kit (7673A), includes six 10 μl 23/42/cone syringes, 4 ml vial with diffusion cap (144/pack), 2 ml screw-top vials with caps (1000/pack), inlet septa (100/pack), vial rack (5/pack).
- Désignation:Basic automatic liquid sampler supply kit