Erlenmeyer Flasks, PETG, Flat Bottom
Supplier:   NEST SCIENTIFIC
Erlenmeyer Flasks, PETG, Flat Bottom
Supplier:  NEST SCIENTIFIC
Certificates
Erlenmeyer Flasks, PETG, Flat Bottom
8 Options Available Below
Product Details & Documents
Ideal for shaker culture applications. PETG Erlenmeyer flasks are manufactured in 100000 grade cleaning room with USP 6 grade raw materials.
  • PETG Bottle and HDPE cap
  • Sterilized by E-Beam
  • Nonpyrogenic
  • Packaged in sterile bags
Vent caps with 0.22 µm hydrophobic filters for gas exchange without contamination.
 : ISO 9001 and 13485 certified.
Product Family Options
With plug seal capped
Product Information
  • Désignation
    Capacité
    Sterility
    Hauteur
    Neck O.D.
    Availability
    Price
  • Conical PETG Erlenmeyer flasks, flat bottom, Bottom Ø: 66 mm, 24 per case
    125 ml
    Sterile
    106.8 mm
    34 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $197.71
    $197.71
    Log in for availability
    Each
    Each - $197.71
    $197.71
    Case of 24
       $197.71
    Each
       $197.71
    Supplier Number: 781001
  • Conical PETG Erlenmeyer flasks, flat bottom, Bottom Ø: 83 mm, 12 per case
    250 ml
    Sterile
    137.8 mm
    34 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $138.20
    $138.20
    Log in for availability
    Each
    Each - $138.20
    $138.20
    Case of 12
       $138.20
    Each
       $138.20
    Supplier Number: 782001
  • Conical PETG Erlenmeyer flasks, flat bottom, Bottom Ø: 101 mm, 12 per case
    500 ml
    Sterile
    175.8 mm
    39 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $186.28
    $186.28
    Log in for availability
    Each
    Each - $186.28
    $186.28
    Case of 12
       $186.28
    Each
       $186.28
    Supplier Number: 783001
  • Conical PETG Erlenmeyer flasks, flat bottom, Bottom Ø: 127 mm, 6 per case
    1000 ml
    Sterile
    213.3 mm
    39 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 6
    Case of 6 - $153.54
    $153.54
    Log in for availability
    Each
    Each - $153.54
    $153.54
    Case of 6
       $153.54
    Each
       $153.54
    Supplier Number: 784001
With vented cap
Product Information
  • Désignation
    Capacité
    Sterility
    Hauteur
    Neck O.D.
    Availability
    Price
  • Conical PETG Erlenmeyer flasks, flat bottom, Bottom Ø: 66 mm, 24 per case
    125 ml
    Sterile
    106.8 mm
    34 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $209.04
    $209.04
    Log in for availability
    Each
    Each - $209.04
    $209.04
    Case of 24
       $209.04
    Each
       $209.04
    Supplier Number: 781011
  • Conical PETG Erlenmeyer flasks, flat bottom, Bottom Ø: 83 mm, 12 per case
    250 ml
    Sterile
    137.8 mm
    34 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $142.44
    $142.44
    Log in for availability
    Each
    Each - $142.44
    $142.44
    Case of 12
       $142.44
    Each
       $142.44
    Supplier Number: 782011
  • Conical PETG Erlenmeyer flasks, flat bottom, Bottom Ø: 101 mm, 12 per case
    500 ml
    Sterile
    175.8 mm
    39 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $195.77
    $195.77
    Log in for availability
    Each
    Each - $195.77
    $195.77
    Case of 12
       $195.77
    Each
       $195.77
    Supplier Number: 783011
  • Conical PETG Erlenmeyer flasks, flat bottom, Bottom Ø: 127 mm, 6 per case
    1000 ml
    Sterile
    213.3 mm
    39 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 6
    Case of 6 - $162.76
    $162.76
    Log in for availability
    Each
    Each - $162.76
    $162.76
    Case of 6
       $162.76
    Each
       $162.76
    Supplier Number: 784011