High Efficiency Erlenmeyer Flasks, Polycarbonate, Flat Bottom
Supplier: NEST SCIENTIFIC
Jump to:
À propos de cet article
High-efficiency, large-volume culture flasks allow cells to show strong viability with large expression amount of proteins in the culture of mammalian cells and insect cells.
- PC bottle, which meets the USP Class 6 Standard, with high transparency, has strong impact resistance, oxidation resistance and can withstand a high temperature up to 121 ℃
- There’s a scale made by injection molding on the flask body to facilitate observing the filled liquid volume
- Sterilized by E-beam, SAL= 10⁻⁶
- Non-pyrogenic, DNase/Rnase free
- Individually packaged in sterile bag
During the culturing process, the use rate of the shaker is significantly increased, and the survival rate and
viability of cells are both dramatically elevated.
8 Options Available Below
Product Details & Documents
High-efficiency, large-volume culture flasks allow cells to show strong viability with large expression amount of proteins in the culture of mammalian cells and insect cells.
- PC bottle, which meets the USP Class 6 Standard, with high transparency, has strong impact resistance, oxidation resistance and can withstand a high temperature up to 121 ℃
- There’s a scale made by injection molding on the flask body to facilitate observing the filled liquid volume
- Sterilized by E-beam, SAL= 10⁻⁶
- Non-pyrogenic, DNase/Rnase free
- Individually packaged in sterile bag
During the culturing process, the use rate of the shaker is significantly increased, and the survival rate and
viability of cells are both dramatically elevated.
Product Family Options
With sealed cap
Informations sur le produit
- DésignationCapacitéSterilityAvailabilityPrice
- PC High efficient Erlenmeyer flask, flat bottom2 LSterileCase of 4$268.40Each$268.40Supplier Number: 785101
- PC High efficient Erlenmeyer flask, flat bottom3 LSterileCase of 4$458.74Each$458.74Supplier Number: 786101
- PC High efficient Erlenmeyer flask, flat bottom, wide mouth3 LSterileCase of 4$458.74Each$458.74Supplier Number: 786501
- PC High efficient Erlenmeyer flask, flat bottom5 LSterileCase of 4$680.66Each$680.66Supplier Number: 787001
With vented cap
Informations sur le produit
- DésignationCapacitéSterilityAvailabilityPrice
- PC High efficient Erlenmeyer flask, flat bottom2 LSterileCase of 4$277.15Each$277.15Supplier Number: 785111
- PC High efficient Erlenmeyer flask, flat bottom3 LSterileCase of 4$464.39Each$464.39Supplier Number: 786111
- PC High efficient Erlenmeyer flask, flat bottom, wide mouth3 LSterileCase of 4$468.72Each$468.72Supplier Number: 786511
- PC High efficient Erlenmeyer flask, flat bottom5 LSterileCase of 4$694.48Each$694.48Supplier Number: 787011