Erlenmeyer Flasks, Polycarbonate, Flat Bottom
Erlenmeyer Flasks, Polycarbonate, Flat Bottom
Erlenmeyer Flasks, Polycarbonate, Flat Bottom
PC Erlenmeyer flasks are manufactured in 100000 grade cleaning room with USP 6 grade raw materials.
  • PC Bottle and HDPE cap
  • Vent caps with 0.2 μm hydrophobic filters for gas exchange without contamination
  • Individually packaged in sterile bags
  • Sterilized by E-beam, SAL= 10⁻⁶
  • Non-pyrogenic, DNase/Rnase free
PC Bottle and HDPE cap, vent caps with 0.2 µm hydrophobic filters for gas exchange without contamination. Ideal for shaker culture applications.
 : ISO 9001 and 13485 certified.
Product Family Options
With sealed cap
  • Désignation
    Capacité
    Sterility
  • Conical Erlenmeyer flasks, flat bottom
    2 L
    Sterile
    Case of 6
    Case of 6 - $402.53
    $402.53
    Each
    Each - $402.53
    $402.53
    Case of 6
       $402.53
    Each
       $402.53
     : 785001
  • Conical Erlenmeyer flasks, flat bottom
    3 L
    Sterile
    Case of 4
    Case of 4 - $471.56
    $471.56
    Each
    Each - $471.56
    $471.56
    Case of 4
       $471.56
    Each
       $471.56
     : 786001
With vented cap
  • Désignation
    Capacité
    Sterility
  • Conical Erlenmeyer flasks, flat bottom
    2 L
    Sterile
    Case of 6
    Case of 6 - $417.59
    $417.59
    Each
    Each - $417.59
    $417.59
    Case of 6
       $417.59
    Each
       $417.59
     : 785011
  • Conical Erlenmeyer flasks, flat bottom
    3 L
    Sterile
    Case of 4
    Case of 4 - $477.19
    $477.19
    Each
    Each - $477.19
    $477.19
    Case of 4
       $477.19
    Each
       $477.19
     : 786011