KIMAX® Culture Tubes, Plain, Reusable, Borosilicate Glass, Kimble Chase
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
À propos de cet article
Tubes feature a fire-polished top rim and uniform wall thickness for maximum heat transfer and chemical resistance.
Made from KIMAX-51® borosilicate glass, these round-bottom culture tubes are supplied without marking area or cap. Designed from ASTM Specification E982, Type IV requirements.
Frequently Bought Together