KIMAX® Centrifuge Tube, Conical-Bottom, Graduated, with Pourout, Kimble Chase
Catalogue # 89002-206
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Volume:50 ml
- Cat. no.:89002-206
- O.D.×L:29×118 mm
- Supplier No.:45186-50
Spécifications
À propos de cet article
Tube has a short tapered bottom and a pour spout.
- Graduated and calibrated to contain
Scale, legend and marking spot are durable white ceramic enamel. Graduated to 10 ml in 0.5 ml increments and from 10 to 50 ml in 1 ml increments.