KIMAX® Short Cone Oil Centrifuge Tube, Kimble Chase
Catalogue # 89002-230
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Volume:100 ml
- Description:KIMAX® Short Cone Oil Centrifuge Tube
- Graduation increments:0.05 mL from 0 to 0.5 mL, 0.1 mL from 0.5 to 2 mL, 0.2 mL from 2 to 3 mL, 0.5 mL from 3 to 5 mL, 1 mL from 5 to 10 mL, 5 mL from 10 to 25 mL, 25 mL from 25 to 50 mL, 50 mL from 50 to 100 mL
- Tolerance:±0.02 mL from 0 to 0.1 mL, ±0.03 mL from 0.1 to 0.3 mL, ±0.05 mL from 0.3 to 0.5 mL, ±0.075 mL from 0.5 to 1 mL, ±0.1 mL from 1 to 1.5 mL, ±0.2 mL from 1.5 to 2 mL, ±0.3 mL from 2 to 3 mL, ±0.5 mL from 3 to 5 mL, ±0.75 mL from 5 to 10 mL, ±1 mL from 10 to 25 mL, ±1.5 from 25 to 100 mL
- Cat. no.:89002-230
- Supplier No.:45243-100
Spécifications
À propos de cet article
Designed for field use in petroleum testing, these KIMAX® tubes are calibrated to contain and tooled to accept a snap cap.
- Dimensions: 45Ext.Ø×165L mm (1³/₄×6").
- Centrifuge tubes are made of borosilicate glass
Scale and legend are durable white ceramic enamel.
Certifications: Referenced in ASTM D4007.