VWR® Peg Racks | Avantor
VWR® Peg Racks
White, 96, 10 - 13 mm
Catalog # 82024-496
Supplier:   VWR International
VWR® Peg Racks
Catalog # 82024-496
Supplier:  VWR International
VWR® Peg Racks
VWR® Peg Racks
White, 96, 10 - 13 mm
Price
64,04 $
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Pack of 2 Case of 8
Product Details & Documents
Ideal for bench work or storage, these polypropylene racks are reinforced with fiberglass to make them strong and versatile.
  • Polypropylene construction
  • Fiberglass reinforcement
  • Autoclavable
Racks feature embossed indexing, ribbed grips and drainage holes. Spacious peg design allows easy viewing of tube contents. Tubes may be placed in the rack upside down for drying. Pegs remain stiff and straight through repeated autoclaving.
Spécifications
  • Nb de places:
    96
  • Color:
    White
  • For Tube O.D.:
    10 - 13 mm