J.T.Baker®, pH Buffer Solutions
About this item
High quality buffer solutions ensure accurate pH measurements time after time.
- Colour coded
- Potassium hydrogen phthalate based pH 4 buffer (red)
- Phosphate based pH 7 buffer (yellow)
- Borate based pH 10 buffer (blue)
Ready-to-use pH buffers are manufactured in large lots to stringent specifications and utilize quality control procedures to reduce lot-to-lot variability. You can eliminate the time and expense of preparation and standardization of solutions, helping streamline high-volume analytical work in applications such as quality control.
Solutions are available in both clear and color-coded varieties.
A certificate is provided to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Certifications: Certified traceable to NIST Standard Reference Buffers.
8 Options Available Below
Product Details & Documents
High quality buffer solutions ensure accurate pH measurements time after time.
- Colour coded
- Potassium hydrogen phthalate based pH 4 buffer (red)
- Phosphate based pH 7 buffer (yellow)
- Borate based pH 10 buffer (blue)
Ready-to-use pH buffers are manufactured in large lots to stringent specifications and utilize quality control procedures to reduce lot-to-lot variability. You can eliminate the time and expense of preparation and standardization of solutions, helping streamline high-volume analytical work in applications such as quality control.
Solutions are available in both clear and color-coded varieties.
A certificate is provided to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Certifications: Certified traceable to NIST Standard Reference Buffers.
Documents
Product Family Options
Product Information
- Type de conditionnementTaillepHColorAvailabilityPrice
- PE bottle500 ml4.00ColorlessDiscontinued-Supplier Number: -Specifications:MOC: Non-Regulated, With InformationMOC: Non-Regulated, With Information
- PE bottle500 ml4.00ColorlessEach(1/Itm/Bx)$116.45Case of 12 (1/ Itm/Bx)$1,073.66Supplier Number: 5606-01Specifications:MOC: Non-Regulated, With InformationMOC: Non-Regulated, With Information
- Cubetainer®4 L4.00ColorlessEach (4l)$409.00Case of 4 (4l)$1,636.00Supplier Number: BAKR5606-03Specifications:MOC: Non-Regulated, With InformationMOC: Non-Regulated, With Information
- PE bottle500 ml4.00RedEach(1/Itm/Bx)$129.60Case of 12 (1/ Itm/Bx)$1,088.91Supplier Number: 5657-01Specifications:MOC: Non-Regulated, With InformationMOC: Non-Regulated, With Information
- PE bottle500 ml10.00BlueEach (500ml)$116.72Case of 12 (500ml)$1,081.38Supplier Number: BAKR5655-01Specifications:MOC: Non-Regulated, With InformationMOC: Non-Regulated, With Information
- Cubetainer®4 L10.00BlueEach (4l)$353.09Case of 4 (4l)$1,138.98Supplier Number: BAKR5655-03Specifications:MOC: Non-Regulated, With InformationMOC: Non-Regulated, With Information
- Supplier Number: BAKR5655-07Specifications:MOC: Non-Regulated, With InformationMOC: Non-Regulated, With Information
- PE bottle500 ml7.00YellowEach (500ml)$116.25Case of 12 (500ml)$1,075.32Supplier Number: BAKR5656-01Specifications:MOC: Non-Regulated, With InformationMOC: Non-Regulated, With Information
- Cubetainer®20 L7.00YellowDiscontinued-Supplier Number: -Specifications:MOC: Non-Regulated, With InformationMOC: Non-Regulated, With Information
- PE bottle500 ml4.00RedDiscontinued-Supplier Number: -Specifications:MOC: Non-Regulated, With InformationMOC: Non-Regulated, With Information
- Cubetainer®4 L4.00RedDiscontinued-Supplier Number: -Specifications:MOC: Non-Regulated, With InformationMOC: Non-Regulated, With Information
- Supplier Number: BAKR5657-07Specifications:MOC: Non-Regulated, With InformationMOC: Non-Regulated, With Information