Synthware Extraction Thimbles, Glass
Supplier: Kemtech America
Jump to:
À propos de cet article
All-glass extraction thimbles permit viewing extraction process and subsequent weighing.
- Heavy wall borosilicate construction
- Coarse or extra coarse porosity available
: Small thimble is used with condenser 60002-198 and extractor 60002-204; Medium thimble is used with condenser 60002-200 and extractors 60002-206 or 60002-208; Large thimble is used with condenser 60002-202 and extractors 60002-210 or 60002-212.
6 Options Available Below
Product Details & Documents
All-glass extraction thimbles permit viewing extraction process and subsequent weighing.
- Heavy wall borosilicate construction
- Coarse or extra coarse porosity available
: Small thimble is used with condenser 60002-198 and extractor 60002-204; Medium thimble is used with condenser 60002-200 and extractors 60002-206 or 60002-208; Large thimble is used with condenser 60002-202 and extractors 60002-210 or 60002-212.
Product Family Options
Informations sur le produit
- FitsO.D.×HPorositéAvailabilityPrice
- Supplier Number: E112585C
- Supplier Number: E112585X
- Supplier Number: E113590C
- Supplier Number: E113590X
- Supplier Number: E114513C
- Supplier Number: E114513X