dNTP Mix, 10 mM, QuantaBio
dNTP Mix 10 mM, 1000 μL
Catalog # CA101414-200
Supplier: Quantabio
Fournisseur #: 95062-01K
Jump to:
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Return Policy
Product Details & Documents
The 10mM dNTP Mix is a solution of high purity deoxyribonucleoside triphosphates that has been functionally qualified for real-time quantitative PCR (qPCR)
- High Purity
- Functionally tested
- Improves dynamic range, sensitivity and precision of qPCR assays
It is suitable for use in conventional end-point PCR, qPCR, first-strand cDNA synthesis, and other applications that require dNTP as substrate. Quality and purity of dNTP can directly affect dynamic range, sensitivity and precision of qPCR assays. The use of qPCR certified dNTPs is a simple but important measure to ensure consistent assay performance.
Spécifications
Spécifications
- Désignation:dNTP Mix 10 mM
- Taille:1000 μL