Kimble® Disposable Plain Conical Centrifuge Tubes with Snap Cap Rim, Glass, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
À propos de cet article
Borosilicate glass (N-51A) conical tubes are constructed with a finished top for polyethylene snap-top closures.
Certifications: Meet Type I, A, or B glass requirements per ASTM E438.
Ordering information: Supplied shrink-wrapped on trays without caps. Order caps separately.
Frequently Bought Together