Alundum® Extraction Thimbles
Alundum® Extraction Thimbles
Alundum® Extraction Thimbles
Certificates
Alundum® Extraction Thimbles
Alundum® Extraction Thimbles
2 Options Available Below
Product Details & Documents
Thimbles remain very constant in weight
Preferable to paper thimbles as they may be used an indefinite number of times and permit the ignition of the extracted residue without removing the material from the thimble. To clean, place in a strong solution of acid for a short time and then ignite to constant weight in the usual way. Available in three degrees of porosity.
Product Family Options
Informations sur le produit
  • Shape
    Hauteur
    Ø ext.
    Porosité
    Availability
    Price
  • Flat
    70 mm
    25 mm
    Medium
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $485.31
    $485.31
    Log in pour la disponibilité
    Pack of 12
    Pack of 12 - $5,823.72
    $5,823.72
    Each
       $485.31
    Pack of 12
       $5,823.72
    Supplier Number: 3254422
  • Flat
    70 mm
    25 mm
    Medium
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 12
    Pack of 12 - $4,259.58
    $4,259.58
    Pack of 12
       $4,259.58
    Supplier Number: 3254422
  • Flat
    80 mm
    20 mm
    Medium
    Discontinued
    Supplier Number: -
  • Round
    80 mm
    25 mm
    Dense
    Discontinued
    Supplier Number: -
  • Round
    100 mm
    34 mm
    Medium
    Discontinued
    Supplier Number: -