PYREX® Extraction Thimbles, with Fritted Disc, Corning
PYREX® Extraction Thimbles, with Fritted Disc, Corning
   Options durables disponibles
Supplier:   Corning
PYREX® Extraction Thimbles, with Fritted Disc, Corning
Supplier:  Corning
   Options durables disponibles
Certificates
PYREX® Extraction Thimbles, with Fritted Disc, Corning
PYREX® Extraction Thimbles, with Fritted Disc, Corning
5 Options Available Below
Product Details & Documents
PYREX®, borosilicate glass, with fritted disc.
  • Resistant to chemical attack
  • Excellent temperature resistance
It is designed for use with soxhlet extractors. It is also useful for chemical applications where it is required to weigh the extracted material.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 89091-272
  • 89091-346
  • 89091-384
  • 27743-120
  • 27743-141
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
Spécifications
Product Family Options
Coarse
Informations sur le produit
  • Taille
    Diameter ×H
    Porosité
    Availability
    Price
  • Small
    25×85 mm
    40-60 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 9
    Case of 9 - 1 150,00 $
    1 150,00 $
    Case of 9
       1 150,00 $
    Supplier Number: 33950-SC
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89091-272
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • Medium
    35×90 mm
    40-60 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 6
    Case of 6 - 851,00 $
    851,00 $
    Case of 6
       851,00 $
    Supplier Number: 33950-MC
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89091-346
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • Large
    45×130 mm
    40-60 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 6
    Case of 6 - 1 070,00 $
    1 070,00 $
    Case of 6
       1 070,00 $
    Supplier Number: 33950-LC
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89091-384
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
Extra Coarse
Informations sur le produit
  • Taille
    Diameter ×H
    Porosité
    Availability
    Price
  • Medium
    35×90 mm
    170-200 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - 162,00 $
    162,00 $
    Case of 1
       162,00 $
    Supplier Number: 33950-MEC
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    27743-120
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • Large
    45×130 mm
    170-200 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - 206,00 $
    206,00 $
    Case of 1
       206,00 $
    Supplier Number: 33950-LEC
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    27743-141
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact