PYREX® Accelerated Modular One-Step™ Extraction Apparatus, Corning | Avantor
PYREX® Accelerated Modular One-Step™ Extraction Apparatus, Corning
   Options durables disponibles
Supplier:   Corning
PYREX® Accelerated Modular One-Step™ Extraction Apparatus, Corning
Supplier:  Corning
   Options durables disponibles
Certificates
PYREX® Accelerated Modular One-Step™ Extraction Apparatus, Corning
9 Options Available Below
Product Details & Documents
This revolutionary modular extractor-concentrator performs extraction, concentration and drying with the same apparatus and is designed for use with solvents of heavier specific gravity than sample being extracted.
Removable Snyder column sidearm allows easy cleaning. Extraction and concentration steps can be performed outside a hood, although the use of a ventilation system is recommended above or near the units in case of a drop in condenser water temperature or loss of condenser cold water supply.
Certifications: Meets EPA test requirements outlined by SW-846 method 3520 and Waste Water method 625 allowed in CLP statement of work OLMO2.0 for semi-volatiles and pesticides.
Ordering information: This apparatus requires a PYREX® concentrator tube (27614-428) and PYREX® Allihn condenser (see 27613-022 series), snyder sidearm, extractor cup and body, O-rings, connectors, membrane and stopcock assembly and membrane support should be ordered separately..
Specifications for Product Family
Spécifications
  • Catalog No:
  • 27614-430
  • 27614-438
  • 27614-440
  • 27614-444
  • 27614-448
  • 27614-432
  • 27614-434
  • 27614-442
  • 27614-424
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
Spécifications
Product Family Options
Replacement Parts
Informations sur le produit
  • Désignation
    Availability
    Price
  • Membrane Assembly, 47 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 20
    Case of 20 - $1,810.00
    $1,810.00
    Case of 20
       $1,810.00
    Supplier Number: 3918-47
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    27614-430
    Environmentally Preferable
    Environmental certification
    Product available on GSA Advantage®
  • Coupler Assembly, Top and Bottom
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 6
    Case of 6 - $1,020.00
    $1,020.00
    Case of 6
       $1,020.00
    Supplier Number: 3915-CAO
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    27614-438
    Environmentally Preferable
    Environmental certification
    Product available on GSA Advantage®
  • Photoceram™ Support for 47 mm Membrane Assembly
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 2
    Case of 2 - $1,100.00
    $1,100.00
    Case of 2
       $1,100.00
    Supplier Number: 3918-47MS
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    27614-440
    Environmentally Preferable
    Environmental certification
    Product available on GSA Advantage®
  • Connector and Washers for Stopcock Assembly
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 6
    Case of 6 - $729.00
    $729.00
    Case of 6
       $729.00
    Supplier Number: 3915-CW
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    27614-444
    Environmentally Preferable
    Environmental certification
    Product available on GSA Advantage®
  • O-Rings, Viton® Coupler
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 24
    Case of 24 - $424.00
    $424.00
    Case of 24
       $424.00
    Supplier Number: 3915-CVO
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    27614-448
    Environmentally Preferable
    Environmental certification
    Product available on GSA Advantage®
  • Extractor Body, Top [ST] 45/50, Side [SJ] 35/25
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $1,180.00
    $1,180.00
    Case of 1
       $1,180.00
    Supplier Number: 3915-MBO
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    27614-432
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • Extractor Cup
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $482.00
    $482.00
    Case of 1
       $482.00
    Supplier Number: 3915-ECO
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    27614-434
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • Stopcock Assembly
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $301.00
    $301.00
    Case of 1
       $301.00
    Supplier Number: 3915-SCA
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    27614-442
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
Informations sur le produit
  • Désignation
    Availability
    Price
  • Complete Apparatus
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $4,120.00
    $4,120.00
    Case of 1
       $4,120.00
    Supplier Number: 3915-C
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    27614-424
    Environmentally Preferable
    Environmental certification
    Product available on GSA Advantage®