ContactUsLinkComponent
Richard-Allan Scientific® Cytoseal™ XYL Mounting Medium, Thermo Scientific
Catalogue # CA48212-196
Fournisseur:  Thermo Scientific
Numéro CAS:  
undefined
Richard-Allan Scientific® Cytoseal™ XYL Mounting Medium, Thermo Scientific
Catalogue # CA48212-196
Fournisseur:  Thermo Scientific
Supplier Number:  8312-4
Numéro CAS:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Spécifications
  • Description:
    Cytoseal™ XYL Mounting Medium
  • Size:
    118.3 mL (4 oz.)
  • Cat. no.:
    CA48212-196
  • Supplier No.:
    8312-4
Spécifications
À propos de cet article
Inhibit the fading or yellowing of stained specimens with specially formulated Thermo Scientific™ Richard-Allan Scientific™ Cytoseal™ XYL mounting media.
  • Xylene-based rapid drying allows microscopic examination of slides soon after application
  • Simple dispensing
This xylene-based mounting medium is made of high-quality synthetic resins and contains an antioxidant to inhibit stain fading and yellowing of specimen. Rapid drying medium allows specimen examination soon after application. Bottle features a twist-open cap for simplified dispensing.