Tissue Freezing Media (TFM™), TBS®, General Data
Supplier: General Data Healthcare
Certificates
About this item
This fast-freezing, cryo-embedding matrix improves specimen handling
Unique formula reduces frozen specimen curling and simplifies pick-up of flat serial sections. Reduced water content minimizes freeze-fractures. Color variety makes differentiating multiple specimens easy. Size: 118mL (4oz.).
Ordering information: Assortment 15146-019 includes one bottle of each color in a wooden rack.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together