Face Shield with Ratchet Headgear, Sellstrom®
Supplier: Sellstrom
Certificates
About this item
Face shield with crown and chin protector offers extra protection from splashes, dust, and other contaminants
- Crown and chin protector offers extra protection from splashes, dust, and other contaminants
- Durable ratchet headgear, turn-of-the-knob adjustability
- Acetate window is 1mm (0.040") thick and measures 17.7Hx48.2Wcm (7x19")
- High-impact sparkguard for extra protection
Note: Face shield should be worn over ANSI-approved protective eyewear.
Durable ratchet headgear allows for maximum comfort with easy turn-of-the-knob adjustability while being worn. Headgear comes with clear window 33007-064. Acetate window is 1mm (0.040") thick and measures 17.7H x 48.2Wcm (7 x 19"). High impact sparkguard for extra protection. Replacement windows available as regular or fog-free.
Frequently Bought Together