KIMAX® Specific Gravity Bottle, Hubbard, Kimble Chase
Catalog # 16620-004
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Capacity:24 ml (0.81 oz.)
- Cat. no.:16620-004
- ST Stopper No.:24/12
- Supplier No.:15110-24
Specifications
About this item
For specific gravity of road oils, road tars, soft tar pitches, and asphalt cements according to ASTM D70, AASHTO T43; drying oils, varnishes, resins, and related materials (ASTM D1963); water and brine (ASTM D1429); and insulating varnishes (ASTM D115)
Stopper is concave on underside and has hole, approx. 1.6 mm in diameter, to allow air to escape.