PYREX® Specific Gravity Bottle, Hubbard-Carmick, Corning
Catalog # 89091-264
Supplier: Corning
Specifications
- Capacity:25 ml (0.85 oz.)
- Dimensions:40Dia. x 57H mm
- Cat. no.:89091-264
- Supplier No.:1620-25
Specifications
About this item
For viscous fluids, semi-solid bitumens, and emulsions according to ASTM D70. Includes NS 24/12 stopper.