Ferrule Kit for GC-ICP-MS Interface 7500
AGG3158-65004
- Description:Ferrule kit
- Cat. no.:AGG3158-65004
- For Use With:For GC-ICP-MS interface 7500
Kit contains:
Reducer ferrule (⅛ to ¹⁄₁₆", 2/pack)
Graphite ferrule (320 µm, int.Ø 0.5 mm, 2/pack)
Vespel-graphite ferrule (¼", 2/pack)
Vespel-graphite ferrule (¹⁄₁₆", 2/pack)