Erlenmeyer Flasks, Closed System | Avantor
Erlenmeyer Flasks, Closed System
Supplier:   NEST SCIENTIFIC
Erlenmeyer Flasks, Closed System
Supplier:  NEST SCIENTIFIC
Certificates
Erlenmeyer Flasks, Closed System
2 Options Available Below
Product Details & Documents
For aseptic transfer of culture medium and cell culture solution.
  • Filter membrane area (0.22 μm): Ø 42 mm
  • Tube material: TPE
One end of the pipeline is connected to the MPC series connector, which increases the convenience and safety in use. The male connector and the sealing cap are tightly connected to prevent accidental disconnection, and the connector can be rotated to reduce pipeline distortion.
Product Family Options
Informations sur le produit
  • Désignation
    Capacité
    Sterility
    Neck I.D.
    Neck O.D.
    Longueur Tubes
    Availability
    Price
  • Erlenmeyer flask with Bi-directional transfer cap, TPE tube, heat seal
    250 ml
    Sterile
    ¹/₈"
    ¹/₄"
    50 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 4
    Case of 4 - $761.65
    $761.65
    Log in pour la disponibilité
    Each
    Each - $761.65
    $761.65
    Case of 4
       $761.65
    Each
       $761.65
    Supplier Number: C30921-AZC050A
  • Erlenmeyer flask with Bi-directional transfer cap, TPE tube, heat seal
    1000 ml
    Sterile
    ¹/₈"
    ¹/₄"
    50 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 4
    Case of 4 - $809.98
    $809.98
    Log in pour la disponibilité
    Each
    Each - $809.98
    $809.98
    Case of 4
       $809.98
    Each
       $809.98
    Supplier Number: C31121-AZC050A