Inoculating Needle/Loops
Supplier: NEST SCIENTIFIC
Jump to:
À propos de cet article
Inoculating needle/loops made up of PP material, hydrophilic.
- Applied in microbiology experiments
- Different colors for identification
- Sterilized by ethylene oxide
- Easy-tear sterile individual packaging
Flexible handles for easier collection and inoculation.
3 Options Available Below
Product Details & Documents
Inoculating needle/loops made up of PP material, hydrophilic.
- Applied in microbiology experiments
- Different colors for identification
- Sterilized by ethylene oxide
- Easy-tear sterile individual packaging
Flexible handles for easier collection and inoculation.
Product Family Options
Informations sur le produit
- DésignationColorPackagingAvailabilityPrice
- Inoculating needleWhite400/pack, 4000/caseCase of 4,000$764.92Each$764.92Supplier Number: 716001
- Inoculating loop, 1 μlBlue400/pack, 4000/caseCase of 4,000$764.92Each$764.92Supplier Number: 717101
- Inoculating loop, 10 μlYellow400/pack, 4000/caseCase of 4,000$771.91Each$771.91Supplier Number: 718201