ContactUsLinkComponent
Sample Loop, for Tekmar AQUATek 90
Catalogue # AG5182-9689
Sample Loop, for Tekmar AQUATek 90
Catalogue # AG5182-9689
Supplier Number:  5182-9689
Spécifications
  • Capacity:
    5 ml
  • Description:
    Sample loop, for Aquatek 70
  • Cat. no.:
    AG5182-9689