Easy-fit ISIS 3 Sample Loops for ICP-MS
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Certificates
About this item
Assembled loops with fittings, ready to install.
- Pre-assembled and secured to prevent kinks
- Compatible with AVS MS
Fixed-volume loops simplify setup of the ISIS 3 with predetermined load, acquisition, and wash out timing. So, no need to manually determine and set the timing using a signal monitor.
Stabilization time: 20 seconds
Frequently Bought Together