VWR® Slide Mailers for 2 Slides, Color-Coded
Supplier: VWR International
Certificates
About this item
With large, easy-to-secure snaps, these mailers provide safe storage of valuable slides.
- Polypropylene construction
- Autoclavable
- Color-coded
These two-place slide mailers securely hold standard 25x75mm (1x3") slides. Mailers are made of polypropylene and are autoclavable. Assorted colors include purple, red, blue, yellow, and orange for easy color coding of samples.
Frequently Bought Together