PYREX® Plain Funnels, Stemless, Corning | Avantor
PYREX® Plain Funnels, Stemless, Corning
   Options durables disponibles
Supplier:   Corning
PYREX® Plain Funnels, Stemless, Corning
Supplier:  Corning
   Options durables disponibles
Certificates
PYREX® Plain Funnels, Stemless, Corning
2 Options Available Below
Product Details & Documents
PYREX®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
These stemless funnels are of molded construction with bottoms ground flat and have a 6 mm opening. For use in the sugar industries.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 89091-526
  • 30272-045
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
Spécifications
Product Family Options
Informations sur le produit
  • Longueur
    Inner Diameter
    Availability
    Price
  • 60 mm
    75 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 48
    Case of 48 - $1,470.00
    $1,470.00
    Case of 48
       $1,470.00
    Supplier Number: 6240-75
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89091-526
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 82 mm
    100 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $42.10
    $42.10
    Case of 1
       $42.10
    Supplier Number: 6240-100
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    30272-045
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact