DrainRack™ Microscope Slide Trays, Simport Scientific
Fournisseur: Simport Scientific
Certificats
À propos de cet article
These rugged trays/drain racks can hold up to 200 microscope slides in 100 individual numbered slots
These microscope slide trays are made of plastic.
All slides are stored upright for easier insertion and removal. Simply tilt them forward or backward with one finger to easily pick up the desired slide. Bar codes can be read without having to remove the slides from the tray. Not autoclavable.
Frequently Bought Together