VWR® Slide Rack for Slide Boxes
Slide rack, 22.8 cm (9"), 71.1 cm (30"), 17.1 cm (6¾")
82024-620
This storage rack makes organization of samples easy by creating a file system for 50- and 100-place slide boxes.
- Clear acrylic construction
- Easy organization
- Color-coded storage system
- Clear view of labels
Clear acrylic construction allows a clear view of the labels on the slide boxes. Place boxes in rows or columns by color to create an easy coding system.
- Profondeur:17.1 cm (6¾")
- Largeur:71.1 cm (30")
- Désignation:Slide rack
- Hauteur:22.8 cm (9")