Control Slides, Azer Scientific
À propos de cet article
Control slides are designed for use in monitoring stain performance in the Pathology lab.
- Permanent positive charge for improved readability
- Control slides with paraffin-embedded human tissue sections
- Ten unstained Unifrost slides
- One stained slide
All slides feature a permanent positive charge that securely anchors all types of sections and improves the quality of preparations and readability
Includes 10 unstained Unifrost microscope slides and one additional slide, which is stained with the proper procedure to demonstrate and validate the expected result.
Azer Scientific's control slides are carefully prepared with paraffin-embedded human tissue sections.
: 11 per pack
15 Options Available Below
Product Details & Documents
Control slides are designed for use in monitoring stain performance in the Pathology lab.
- Permanent positive charge for improved readability
- Control slides with paraffin-embedded human tissue sections
- Ten unstained Unifrost slides
- One stained slide
All slides feature a permanent positive charge that securely anchors all types of sections and improves the quality of preparations and readability
Includes 10 unstained Unifrost microscope slides and one additional slide, which is stained with the proper procedure to demonstrate and validate the expected result.
Azer Scientific's control slides are carefully prepared with paraffin-embedded human tissue sections.
: 11 per pack
Product Family Options
Informations sur le produit
- DésignationL×lAvailabilityPrice
- Control slides, Alcian blue75×25 mm (3×1")Log in pour la disponibilitéLog in pour la disponibilitéPack of 10Pack of 10 - $230.04$230.04Prix promotionnel pour les nouveaux clientsPack of 10$230.04Supplier Number: ESCS-104
- Control slides, Amyloid75×25 mm (3×1")Log in pour la disponibilitéLog in pour la disponibilitéPack of 10Pack of 10 - $164.35$164.35Prix promotionnel pour les nouveaux clientsPack of 10$164.35Supplier Number: ESCS-106
- Control slides, Argentaffin75×25 mm (3×1")Discontinued-Supplier Number: ESCS-107
- Control slides, Bile75×25 mm (3×1")Log in pour la disponibilitéLog in pour la disponibilitéPack of 10Pack of 10 - $158.04$158.04Prix promotionnel pour les nouveaux clientsPack of 10$158.04Supplier Number: ESCS-109
- Control slides, Elastic tissue75×25 mm (3×1")Log in pour la disponibilitéLog in pour la disponibilitéPack of 10Pack of 10 - $173.53$173.53Prix promotionnel pour les nouveaux clientsPack of 10$173.53Supplier Number: ESCS-111
- Control slides, Fungi-gms75×25 mm (3×1")Discontinued-Supplier Number: ESCS-114
- Control slides, Fungi-pas75×25 mm (3×1")Log in pour la disponibilitéLog in pour la disponibilitéPack of 10Pack of 10 - $164.19$164.19Prix promotionnel pour les nouveaux clientsPack of 10$164.19Supplier Number: ESCS-113
- Control slides, Giemsa – for mast cells75×25 mm (3×1")Log in pour la disponibilitéLog in pour la disponibilitéPack of 10Pack of 10 - $166.36$166.36Prix promotionnel pour les nouveaux clientsPack of 10$166.36Supplier Number: ESCS-115
- Control slides, Glycogen75×25 mm (3×1")Log in pour la disponibilitéLog in pour la disponibilitéPack of 10Pack of 10 - $165.61$165.61Prix promotionnel pour les nouveaux clientsPack of 10$165.61Supplier Number: ESCS-116
- Control slides, Gram organism75×25 mm (3×1")Log in pour la disponibilitéLog in pour la disponibilitéPack of 10Pack of 10 - $178.58$178.58Prix promotionnel pour les nouveaux clientsPack of 10$178.58Supplier Number: ESCS-117
- Control slides, Helicobacter75×25 mm (3×1")Log in pour la disponibilitéLog in pour la disponibilitéPack of 10Pack of 10 - $333.27$333.27Prix promotionnel pour les nouveaux clientsPack of 10$333.27Supplier Number: ESCS-130
- Control slides, Iron75×25 mm (3×1")Log in pour la disponibilitéLog in pour la disponibilitéPack of 10Pack of 10 - $185.70$185.70Prix promotionnel pour les nouveaux clientsPack of 10$185.70Supplier Number: ESCS-118
- Control slides, Massons75×25 mm (3×1")Log in pour la disponibilitéLog in pour la disponibilitéPack of 10Pack of 10 - $173.53$173.53Prix promotionnel pour les nouveaux clientsPack of 10$173.53Supplier Number: ESCS-121
- Control slides, Melanin75×25 mm (3×1")Log in pour la disponibilitéLog in pour la disponibilitéPack of 10Pack of 10 - $164.19$164.19Prix promotionnel pour les nouveaux clientsPack of 10$164.19Supplier Number: ESCS-122
- Control slides, Mucicarmine75×25 mm (3×1")Log in pour la disponibilitéLog in pour la disponibilitéPack of 10Pack of 10 - $158.04$158.04Prix promotionnel pour les nouveaux clientsPack of 10$158.04Supplier Number: ESCS-124
- Control slides, Periodic acid75×25 mm (3×1")Log in pour la disponibilitéLog in pour la disponibilitéPack of 10Pack of 10 - $164.44$164.44Prix promotionnel pour les nouveaux clientsPack of 10$164.44Supplier Number: ESCS-125
- Control slides, Ptah75×25 mm (3×1")Discontinued-Supplier Number: ESCS-126
- Control slides, Reticulum75×25 mm (3×1")Discontinued-Supplier Number: ESCS-127
- Control slides, Trichrome75×25 mm (3×1")Log in pour la disponibilitéLog in pour la disponibilitéPack of 10Pack of 10 - $166.60$166.60Prix promotionnel pour les nouveaux clientsPack of 10$166.60Supplier Number: ESCS-129