Histology Control Plus Microscope Slides, Erie Scientific
Ideal for staining procedures that require a control, these frosted slides feature spaces for both the patient sample and the control material
Slides may be used in histology, cytology, or microbiology applications. Slides are designed to ensure that tissue samples remain on the slide. A red or gold box identifies the control section. A red control box on the back surface identifies slides intended for use in automated instruments. Plus Gold control slides are recommended for hard-to-hold sections.
Ideal for staining procedures that require a control, these frosted slides feature spaces for both the patient sample and the control material
Slides may be used in histology, cytology, or microbiology applications. Slides are designed to ensure that tissue samples remain on the slide. A red or gold box identifies the control section. A red control box on the back surface identifies slides intended for use in automated instruments. Plus Gold control slides are recommended for hard-to-hold sections.
Product Family Options
- DésignationL×l
- Superfrost™ Plus, red control box on back75×25 mm-: -
- Superfrost™ Plus, extended 1" white tab75×25 mmPack of 72$127.41Case of 1440$2,548.16: ES-334PLUS-001
- Superfrost™ Plus Gold, special gold adhesion coating75×25 mmPack of 72$237.54Case of 1440$4,750.88: ES-3012-GLPLUS-001
- -: -